Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200647

Sigma-Aldrich

Monoclonal Anti-Strumpellin antibody produced in mouse

clone St-1, purified from hybridoma cell culture

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$530.00

$530.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$530.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$530.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

St-1, monoclonal

form

buffered aqueous solution

mol wt

antigen ~120 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

flow cytometry: 10-20 μg/test using using HeLa cells.
immunoblotting: 2.5-5 μg/mL
immunoprecipitation (IP): 5-10 μg using using lysates of rat A10 cells.

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KIAA0196(9897)

General description

Monoclonal Anti-Strumpellin (mouse IgG1 isotype) is derived from the hybridoma St-1 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide corresponding to an internal region of human Strumpellin, conjugated to KLH.
The gene KIAA0196 (strumpellin) is mapped to human chromosome 8q24. It is a widely expressed gene and the protein localizes in the cytoplasm and endoplasmic reticulum.

Immunogen

synthetic peptide corresponding to an internal region of human Strumpellin.

Application

Monoclonal Anti-Strumpellin antibody has been used in
  • immunoblotting
  • immunoprecipitation
  • flow cytometry

Biochem/physiol Actions

KIAA0196 (strumpellin) is involved in the WASH (WASP and Scar homologue) complex, an actin-regulating complex. It works as an interlink between actin regulation and endosomal membrane dynamics. Mutations in KIAA0196 are associated with spastic paraplegia and RSS (Ritscher-Schinzel syndrome).
Strumpellin, causes hereditary spastic paraplegia, a progressive neurodegenerative disorder clinically characterized by central motor system deficits leading to spastic paraparesis of the lower limbs. Strumpellin presence was observed in protein aggregate diseases affecting striated muscles and the Central nervous system (CNS).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Exome sequencing expands the mutational spectrum of SPG8 in a family with spasticity responsive to L-DOPA treatment.
Conceição Bettencourt et al.
Journal of neurology, 260(9), 2414-2416 (2013-07-25)
A novel KIAA0196 (SPG8) mutation in a Chinese family with spastic paraplegia.
Xianling Wang et al.
Chinese medical journal, 127(10), 1987-1989 (2014-05-16)
Alison M Elliott et al.
Journal of medical genetics, 50(12), 819-822 (2013-09-26)
Ritscher-Schinzel syndrome (RSS) is a clinically heterogeneous disorder characterised by distinctive craniofacial features in addition to cerebellar and cardiac anomalies. It has been described in different populations and is presumed to follow autosomal recessive inheritance. In an effort to identify
Caroline Freeman et al.
Biochimica et biophysica acta, 1832(1), 160-173 (2012-10-23)
Mutations in the gene encoding strumpellin cause autosomal dominant hereditary spastic paraplegia (HSP), in which there is degeneration of corticospinal tract axons. Strumpellin is a component of the WASH complex, an actin-regulating complex that is recruited to endosomes by interactions
Strumpellin is a novel valosin-containing protein binding partner linking hereditary spastic paraplegia to protein aggregation diseases
Clemen C S, et al.
Brain, 133(10), 2920-2941 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service