Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200550

Sigma-Aldrich

Anti-Strumpellin (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-WASH complex subunit strumpellin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~110 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 5-10 μg using lysates of human DU-145 cells.
western blot: 5-10 μg/mL using whole extracts of human PC-3 cells.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Strumpellin is a component of the Wiskott−Aldrich syndrome protein and scar homolog (WASH) multiprotein complex. It is encoded by Wiskott−Aldrich syndrome protein and scar homolog complex subunit 5 (WASHC5) gene. WASHC5 gene is located on the human chromosome at 8q24.13.

Specificity

Anti-Strumpellin (C-terminal) recognizes human Strumpellin.

Immunogen

synthetic peptide corresponding to the C-terminal region of human Strumpellin, conjugated to KLH. The corresponding sequence is identical in mouse and rat.

Application

Anti-Strumpellin (C-terminal) antibody produced in rabbit may be used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

Strumpellin being a component of Wiskott−Aldrich syndrome protein and scar homolog complex (WASH) multiprotein complexis involved in regulating actin. Mutations in the gene encoding strumpellin, KIAA0196 is associated with hereditary spastic paraplegia, a progressive neurodegenerative disorder. Strumpellin is present in protein aggregate diseases affecting striated muscles and the central nervous system (CNS).

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °Cfor up to one month. For extended storage freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of KIAA0196 as a novel susceptibility gene for myofibril structural disorganization in cardiac development
Bu H, et al.
International Journal of Cardiology, 133(10), 2920-2941 (2020)
Strumpellin is a novel valosin-containing protein binding partner linking hereditary spastic paraplegia to protein aggregation diseases
Clemen C S, et al.
Brain, 133(10), 2920-2941 (2010)
The hereditary spastic paraplegia protein strumpellin: characterisation in neurons and of the effect of disease mutations on WASH complex assembly and function
Freeman C, et al.
Biochimica et Biophysica Acta (BBA)-Molecular Basis of Disease, 1832(1), 160-173 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service