Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB4100500

Sigma-Aldrich

Monoclonal Anti-APP antibody produced in mouse

clone APP-B434, culture supernatant

Synonym(s):

Anti-AAA, Anti-ABETA, Anti-ABPP, Anti-AD1, Anti-APPI

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

APP-B434, monoclonal

form

aqueous solution

mol wt

antigen predicted mol wt 60.6 kDa

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... APP(351)

Immunogen

APP purified recombinant protein

Physical form

Solution in Dulbecco′s Modified Eagle′s Medium with 10% fetal calf serum and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service