Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2702177

Sigma-Aldrich

Anti-XPC antibody produced in rabbit

Synonym(s):

RAD4, XP3, XPCC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

human

concentration

0.5 mg/mL

technique(s)

western blot: 1:500-1:1000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... XPC(7508)

General description

XPC (xeroderma pigmentosum group C) is mapped to human chromosome 3p25.1. It has 18 exons and codes for a constituent of the nucleotide excision repair (NER).

Immunogen

Peptide conjugated to a carrier protein

Application

For WB: Load 40ug cell lysate/lane. Use at a dilution of 1:500-1:1000. Optimal dilutions/concentratons should be determined by the researcher.

Biochem/physiol Actions

Human XPC (Xeroderma pigmentosum group C) is a member of a family of proteins that has been shown to be involved in the repair of DNA via the nucleotide excision repair (NER) pathway. Specifically, XPC is believed to be a part of a heteromeric protein complex that is involved in the recognition of the DNA lesions during global genomic repair but not transcription-coupled repair.
XPC (xeroderma pigmentosum group C) participates in repair reaction. XPC protein defends against cancer. XPC plays a major role in the early stages of global genome nucleotide excision repair (NER).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Phosphate-buffered saline, pH 7.2, containing no preservatives

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modulation of repair of ultraviolet damage in the host-cell reactivation assay by polymorphic XPC and XPD/ERCC2 genotypes.
Qiao Y, et al.
Carcinogenesis, 23(2), 295-299 (2002)
Lack of Associations between XPC Gene Polymorphisms and Neuroblastoma Susceptibility in a Chinese Population.
Jintao Z, et al.
BioMed Research International, 2016, 2932049-2932049 (2016)
Xeroderma pigmentosum group C protein interacts with histones: regulation by acetylated states of histone H3.
Kakumu E, et al.
Genes Cells, 22(3), 310-327 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service