Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB2103595

Sigma-Aldrich

Anti-COQ2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CL640, Anti-FLJ26072

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

42 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... COQ2(27235)

Immunogen

Synthetic peptide directed towards the middle region of human COQ2

Biochem/physiol Actions

COQ2 catalyzes the prenylation of para-hydroxybenzoate (PHB) with an all-trans polyprenyl group. COQ2 mediates the second step in the final reaction sequence of coenzyme Q (CoQ) biosynthesis, which is the condensation of the polyisoprenoid side chain with PHB.CoQ (ubiquinone) serves as a redox carrier in the mitochondrial respiratory chain and is a lipid-soluble antioxidant. COQ2, or parahydroxybenzoate-polyprenyltransferase (EC 2.5.1.39), catalyzes one of the final reactions in the biosynthesis of CoQ, the prenylation of parahydroxybenzoate with an all-trans polyprenyl group (Forsgren et al., 2004 [PubMed 15153069]).[supplied by OMIM]. PRIMARYREFSEQ_SPAN PRIMARY_IDENTIFIER PRIMARY_SPAN COMP 1-90 BC020728.1 1-90 91-96 BG699931.1 80-85 97-789 BC020728.1 97-789 790-792 BQ363759.1 169-171 793-1535 BC020728.1 793-1535 1536-1554 BC008804.2 1496-1514

Sequence

Synthetic peptide located within the following region: FSGVMWTLIYDTIYAHQDKRDDVLIGLKSTALRFGENTKPWLSGFSVAML

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service