Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1411452

Sigma-Aldrich

Anti-SEMA6D antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ11598, KIAA1479

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 54.2 kDa

species reactivity

human, mouse

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SEMA6D(80031)

General description

Semaphorins are a large family, including both secreted and membrane associated proteins, many of which have been implicated as inhibitors or chemorepellents in axon pathfinding, fasciculation and branching, and target selection. All semaphorins possess a semaphorin (Sema) domain and a PSI domain (found in plexins, semaphorins and integrins) in the N-terminal extracellular portion. Additional sequence motifs C-terminal to the semaphorin domain allow classification into distinct subfamilies. Results demonstrate that transmembrane semaphorins, like the secreted ones, can act as repulsive axon guidance cues. This gene encodes a class 6 vertebrate transmembrane semaphorin that demonstrates alternative splicing. Six transcript variants have been identified and expression of the distinct encoded isoforms is thought to be regulated in a tissue- and development-dependent manner. (provided by RefSeq)

Immunogen

SEMA6D (NP_079242.2, 1 a.a. ~ 476 a.a) full-length human protein.

Sequence
MRVFLLCAYILLLMVSQLRAVSFPEDDEPLNTVDYHYSRQYPVFRGRPSGNESQHRLDFQLMLKIRDTLYIAGRDQVYTVNLNEMPKTEVIPNKKLTWRSRQQDRENCAMKGKHKDECHNFIKVFVPRNDEMVFVCGTNAFNPMCRYYRLSTLEYDGEEISGLARCPFDARQTNVALFADGKLYSATVADFLASDAVIYRSMGDGSALRTIKYDSKWIKEPHFLHAIEYGNYVYFFFREIAVEHNNLGKAVYSRVARICKNDMGGSQRVLEKHWTSFLKARLNCSVPGDSFFYFDVLQSITDIIQINGIPTVVGVFTTQLNSIPGSAVCAFSMDDIEKVFKGRFKEQKTPDSVWTAVPEDKVPKPRPGCCAKHGLAEAYKTSIDFPDETLSFIKSHPLMDSAVPPIADEPWFTKTRVRYRLTAISVDHSAGPYQNYTVIFVGSEAGMVLKVLAKTSPFSLNDSVLLEEIEAYNHAK

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service