Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300090

Sigma-Aldrich

Anti-Mouse Gpr81-s296 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-FKSG80, Anti-G-protein coupled receptor 104, Anti-G-protein coupled receptor 81, Anti-GPR104, Anti-GPR81

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. Northern analyses revealed GPR81 expression in the pituitary.

Immunogen

GPR81 (NP_780729, 276-329)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the region of mouse GPR81-S296.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rochelle A Peterson et al.
Frontiers in cellular neuroscience, 14, 215-215 (2020-08-28)
A class of Group III muscle afferent neurons has branching sensory terminals in the connective tissue between layers of mouse abdominal muscles ("CT3 muscle afferents"). These sensory endings are both mechanosensitive and metabosensitive. In the present study, responses of CT3
Cecilie Morland et al.
Nature communications, 8, 15557-15557 (2017-05-24)
Physical exercise can improve brain function and delay neurodegeneration; however, the initial signal from muscle to brain is unknown. Here we show that the lactate receptor (HCAR1) is highly enriched in pial fibroblast-like cells that line the vessels supplying blood
Lili Jin et al.
Molecular medicine reports, 26(2) (2022-07-02)
Under aerobic conditions, the preferential use of anaerobic glycolysis by tumour cells leads to a high level of lactate accumulation in tumour microenvironment. Lactate acts not only as a cellular energy source but also as a signalling molecule that regulates
Rupali Vohra et al.
Investigative ophthalmology & visual science, 60(4), 999-1008 (2019-03-19)
Besides being actively metabolized, lactate may also function as a signaling molecule by activation of the G-protein-coupled receptor 81 (GPR81). Thus, we aimed to characterize the metabolic effects of GPR81 activation in Müller cells. Primary Müller cells from mice were
Mohammad Harun-Or-Rashid et al.
Journal of neuroinflammation, 15(1), 313-313 (2018-11-15)
Glaucoma is a chronic degenerative disease for which inflammation is considered to play a pivotal role in the pathogenesis and progression. In this study, we examined the impact of a ketogenic diet on the inflammation evident in glaucoma as a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service