S4576
CleanCut Protease from Staphylococcus aureus
500-1000 units/mg protein
Synonym(s):
SplB Serine Protease
About This Item
biological source
Staphylococcus aureus
assay
95% (SDS-PAGE)
form
lyophilized powder
specific activity
500-1000 units/mg protein
Storage temp.
−20°C
Application
Unit Definition
Physical form
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service