Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S1562

Sigma-Aldrich

Anti-SSeCKS antibody produced in sheep

IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
$526.00

$526.00


Check Cart for Availability


Select a Size

Change View
250 μG
$526.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$526.00


Check Cart for Availability

biological source

sheep

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat

technique(s)

immunoprecipitation (IP): 2-5 μg/mL
western blot (chemiluminescent): 2-5 μg/mL using an extract of rat aortic smooth muscle

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

rat SSeCKS.

Physical form

Solution in phosphate buffered saline with 0.08% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hua Wei et al.
Neurochemical research, 44(4), 839-848 (2019-02-02)
Astrocytes migration is essential in the formation of the glial scar during the injury response process of the central nervous system (CNS) especially during inflammation. Integrin β1 is part of the extracellular matrix receptors in the CNS and it has

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service