Recommended Products
greener alternative product characteristics
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
greener alternative category
shipped in
ambient
storage temp.
room temp
General description
- Isolate true total RNA including siRNA and microRNA
- No phenol extractions
- Purify high-quality RNA in 40 minutes
- Isolate total RNA from a wide variety of sources
Application
- Quantitative, real-time RT-PCR for both large mRNA and small RNA including miRNA
- RT-PCR for both large mRNA and small RNA including miRNA
- Expression profiling
- Next Generation Sequencing for RNA and miRNA
- miRNA from plasma for discovery
- microRNA cloning and amplification
- PCR-based virus detection
- PCR-based viable bacteria detection
- Northern blotting
- RNase protection
- Primer extension
- miRNA array
- RNA-Seq
- Nanostring
- Fluidigm
- Droplet/Digital PCR
Features and Benefits
- Isolate a diversity of RNA species - All RNA species can be isolated, from large mRNA and ribosomal RNA down to microRNA (miRNA) and small interfering RNA (siRNA)
- Fast and easy processing - Rapid spin-column format allows for the processing of 4 samples in under 40 minutes.
- No phenol:chloroform extractions – Total RNA is isolated without the use of harmful chemicals such as phenol or chloroform.
- Isolate total RNA from a wide variety of sources – Total RNA can be isolated from cultured animal cells, tissue samples, blood, bacteria, yeast, fungi, plants and viruses using this kit.
- Isolate inhibitor-free RNA suitable for downstream applications - Purified RNA can be used in a number of downstream applications including real-time PCR, reverse transcription PCR, Northern blotting, RNase protection and primer extension, and expression array analysis requiring the use of intact RNA
- Isolate a diversity of RNA species - All RNA species can be isolated, from large mRNA and ribosomal RNA down to microRNA (miRNA) and small interfering RNA (siRNA)
- Fast and easy processing - Rapid spin-column format allows for the processing of 4 samples in under 40 minutes.
- No phenol:chloroform extractions – Total RNA is isolated without the use of harmful chemicals such as phenol or chloroform.
- Isolate total RNA from a wide variety of sources – Total RNA can be isolated from cultured animal cells, tissue samples, blood, bacteria, yeast, fungi, plants and viruses using this kit.
- Isolate inhibitor-free RNA suitable for downstream applications - Purified RNA can be used in a number of downstream applications including real-time PCR, reverse transcription PCR, Northern blotting, RNase protection and primer extension, and expression array analysis requiring the use of intact RNA
Unit Definition
Other Notes
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1C
supp_hazards
Storage Class
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service