Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PCG3005

Sigma-Aldrich

TruPAGE DTT Sample Reducer

10 ×

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201
NACRES:
NB.22

concentration

10 ×

compatibility

for use with Sigma-Aldrich TruPAGE Precast Gels

storage temp.

2-8°C

Features and Benefits

TruPAGE DTT Sample Reducer should be utilized when performing reducing SDS-PAGE. The Sample Reducer reduces disulfide bonds to help denature protein samples. TruPAGE running buffers are non-reducing, and the cathode chamber should be supplemented with a 1X working concentration of TruPAGE Antioxidant to prevent protein reoxidation and refolding during electrophoresis.

Legal Information

TruPAGE is a trademark of Sigma-Aldrich Co. LLC

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minzi Deng et al.
Experimental cell research, 395(1), 112175-112175 (2020-07-18)
Autophagy is a basic catabolic response that eukaryotic cells use to degrade unnecessary or dysfunctional cellular components in an orderly and regulated manner. It plays important roles in maintaining cellular homeostasis, energy homeostasis, response to environmental stimuli, and the development

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service