Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P9622

SAFC

Pristane

synthetic, liquid, sterile-filtered, BioReagent

Synonym(s):

2,6,10,14-Tetramethylpentadecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CH(CH2)3CH(CH3)(CH2)3CH(CH3)(CH2)3CH(CH3)2
CAS Number:
Molecular Weight:
268.52
Beilstein/REAXYS Number:
1720538
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.75

biological source

synthetic

Quality Level

sterility

sterile-filtered

product line

BioReagent

assay

≥95%

form

liquid

impurities

endotoxin, tested

refractive index

n20/D 1.438 (lit.)

bp

68 °C/0.001 mmHg (lit.)

density

0.783 g/mL at 20 °C (lit.)

SMILES string

CC(C)CCCC(C)CCCC(C)CCCC(C)C

InChI

1S/C19H40/c1-16(2)10-7-12-18(5)14-9-15-19(6)13-8-11-17(3)4/h16-19H,7-15H2,1-6H3

InChI key

XOJVVFBFDXDTEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pristane is a natural saturated terpenoid alkane used in research on the pathogenesis of autoimmune diseases such as arthritis and lupus.

application

Used to induce disease models for study including arthritis as a non-antigenic arthritogenic adjuvant and plasmacytomas.

Packaging

Packaged in flame sealed ampules.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

>230.0 °F - closed cup

flash_point_c

> 110 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen Liu et al.
Journal of autoimmunity, 113, 102498-102498 (2020-06-17)
E proteins, a subset of basic helix-loop-helix (bHLH) proteins, are transcription activators and their functions are inhibited by DNA-binding inhibitor (Id) 1-4. Studies have shown that Treg levels are decreased in Id3 knockout mice. Mice over-expressing Id1 in CD4 T
D A C Botte et al.
Clinical and experimental immunology, 177(2), 381-390 (2014-03-29)
Alpha-melanocyte stimulating hormone (α-MSH) is a neuropeptide exhibiting anti-inflammatory activity in experimental models of autoimmune diseases. However, no studies thus far have examined the effects of α-MSH on systemic lupus erythematosus (SLE). This study aimed to determine the effects of
Nicholas C Van de Velde et al.
Immunology letters, 130(1-2), 82-88 (2009-12-17)
The major human Fc receptor, huFcgammaRIIa, is implicated in the development of autoimmune arthritis in humans but until recently has not been studied in mouse models. We evaluated potential roles of FcgammaRIIa by using transgenic mice expressing the receptor. We
Yiqin Shi et al.
Journal of immunology (Baltimore, Md. : 1950), 193(10), 5129-5139 (2014-10-05)
Diffuse pulmonary hemorrhage (DPH) is an uncommon but critical complication of systemic lupus erythematosus. Peritoneal administration of 2,6,10,14-tetramethylpentadecane (pristane) can recapitulate a lupus-like syndrome in mice, which can develop into DPH within a few weeks, especially in C57BL/6 mice. Mac-1
Sae Won Kim et al.
Oncotarget, 5(5), 1226-1240 (2014-04-10)
Identification of novel biomarkers for tumor-initiating cells (TICs) is of critical importance for developing diagnostic and therapeutic strategies against cancers. Here we identified the role of KIAA1114, a full-length translational product of the trophinin gene, as a distinctive marker for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service