Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P249

Sigma-Aldrich

Anti-Peroxisome Proliferator Activated Receptor antibody produced in rabbit

whole antiserum, buffered aqueous solution

Synonym(s):

Anti-PPAR

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, rat

technique(s)

immunoprecipitation (IP): suitable
western blot: 1:2,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

mouse ... Ppara(19013)
rat ... Ppara(25747)

Immunogen

synthetic peptide corresponding to amino acids 484-498 of the C-terminal region of PPARγ2 isoform. The sequence is highly conserved in PPARγ1, PPARα and PPARβ.

Physical form

Solution in phosphate buffered saline and contains 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service