Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

O0511

Sigma-Aldrich

Octyl−Sepharose 4 Fast Flow

Synonym(s):

Octyl–Agarose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

extent of labeling

~5 μmol per mL

Quality Level

matrix

agarose, 4% cross-linked

matrix activation

epichlorohydrin

matrix attachment

ether

matrix spacer

3 atoms

particle size

45-165 μm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

O0511-200ML′s updated product number is GE17-0946-02

Application

Octyl Sepharose is used in hydrophobic interaction media, resins and separation media. Octyl Sepharose has been used to determine the population of Acinetobacter species in sewage treatment plants.

Physical form

Suspension in 20% ethanol

Legal Information

Sepharose is a trademark of Cytiva

replaced by

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

57.2 °F - closed cup

flash_point_c

14.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P N Ward et al.
Infection and immunity, 66(12), 5636-5642 (1998-11-24)
Pasteurella multocida toxin (PMT) is a potent mitogen that also affects bone resorption. PMT acts intracellularly and is therefore postulated to have several domains involved in different aspects of its function. The toxin contains eight cysteine residues. Mutants with individual
Irina Kostareva et al.
Journal of chromatography. A, 1177(2), 254-264 (2007-11-06)
A heteropolymer (HP) is a unique dual antibody conjugate composed of specific, chemically cross-linked monoclonal antibodies (mAbs). In this study we have demonstrated that HPs can be purified using hydrophobic interaction chromatography (HIC). Two propyl HIC resins; [PolyPropyl A and
Yuko Cho et al.
Biochimica et biophysica acta, 1784(9), 1277-1285 (2008-07-05)
The Japanese bivalve Peronidia venulosa contains paralytic shellfish toxin (PST)-transforming enzymes that convert the weakly toxic C-toxins to the more potent decarbamoyl toxins. The enzyme was purified 154-fold with a yield of 0.26% and was named sulfocarbamoylase I. It was
Florence K Gleason et al.
Journal of bacteriology, 184(23), 6544-6550 (2002-11-12)
The gene for ribonucleotide reductase from Anabaena sp. strain PCC 7120 was identified and expressed in Escherichia coli. This gene codes for a 1,172-amino-acid protein that contains a 407-amino-acid intein. The intein splices itself from the protein when it is
Aviv Duek et al.
Water research, 46(8), 2505-2514 (2012-01-24)
Microorganisms are retained by ultrafiltration (UF) membranes mainly due to size exclusion. The sizes of viruses and membrane pores are close to each other and retention of viruses can be guaranteed only if the precise pore diameter is known. Unfortunately

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service