Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

MAK362

Sigma-Aldrich

Cardiolipin Assay Kit

sufficient for 100 fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

usage

sufficient for 100 fluorometric tests

detection method

fluorometric

storage temp.

−20°C

General description

Cardiolipin (CL) is a very important phospholipid present in the inner mitochondrial membrane and constitutes about 20% of its total lipid composition. It has a dimeric structure comprised of two phosphatidic acid residues linked by a glycerol bridge. It is essential for several mitochondrial functions such as maintaining activity of the electron transport chain complexes, and other mitochondrial processes including biogenesis, fission, fusion and protein transport. It is involved in apoptosis where it helps alter the mitochondrial membrane structure and aids in the release of cytochrome c. Conditions such as diabetes and heart failure are linked to changes in levels of cardiolipin and it is also depleted during aging. Exposure to toxicants like cigarette smoke and organophosphates may cause an alteration in cardiolipin levels and composition, thus adversely affecting health.

Suitability

Suitable for the measurement of cardiolipin content in cell lysates and isolated mitochondria.

Principle

The Cardiolipin Assay Kit is a fluorometric assay that makes use of a proprietary probe that fluoresces on association with cardiolipin but not with any other lipids such as phosphatidylcholine and sphingomyelin, making it highly specific. Fluorescence can be recorded at ex = 340 nm/em = 480 nm. The kit includes purified cardiolipin as standard and can detect as low as 0.2 nmol of cardiolipin.

pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

49.5 °F

flash_point_c

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Does digitonin have any negative effect on cardiolipin when assayed using the MAK362?

    1 answer
    1. The use of a digitonin isolated sample with the MAK362 should not pose any issues.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service