Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK288

Sigma-Aldrich

Purine Nucleoside Phosphorylase Activity Assay Kit (Fluorometric)

sufficient for 100 fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

detection method

fluorometric

relevant disease(s)

immunological diseases; neurological disorders; hematological disorder

storage temp.

−20°C

General description

Purine Nucleoside Phosphorylase (PNP) (E.C. 2.4.2.1.) is an enzyme involved in purine metabolism and it catalyzes the cleavage of the glycosidic bond of ribo- or deoxyribonucleosides, in the presence of inorganic phosphate as a second substrate, to generate the purine base and ribose-1-phosphate or deoxyribose-1-phosphate. It is one of the enzymes of the nucleotide salvage pathways that allows the cell to produce nucleotide monophosphates when the de novo synthesis pathway has been interrupted or is non-existent (as is the case in the brain). PNP deficiency leads to severe combined immunodeficiency and also leads to hypouricemia and reduced urinary excretion of uric acid.[1]

Suitability

Suitable for the detection of Purine Nucleoside Phosphorylase activity in cell and tissue lysate or purified recombinant protein

Principle

In this Purine Nucleoside Phosphorylase Activity Assay, hypoxanthine formed from the breakdown of inosine is detected via a multi-step reaction, resulting in the generation of an intermediate that reacts with the PNP Probe. The fluorescent product is measured at λex = 535nm/λem = 587nm. Limit of quantification is 0.005 μU of recombinant purine nucleoside phosphorylase.

pictograms

Health hazardCorrosion

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Resp. Sens. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Disorders of purine and pyrimidine metabolism.
Nyhan W L, et al.
Molecular Genetics and Metabolism, 86(1), 25-33 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service