Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MAK198

Sigma-Aldrich

2-Phosphoglycerate Assay Kit

Sufficient for 100 Colorimetric or Fluorometric tests

Synonym(s):

2-PG Assay Kit, 2PG Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.84

detection method

colorimetric
fluorometric

storage temp.

−20°C

General description

2-Phosphoglycerate (2PG) is an important intermediate in the glycolysis pathway, a critical mechanism of ATP generation. It is converted to phosphoenol pyruvate (PEP), catalyzed by the enzyme enolase. The consumption of glucose and the rate of glycolysis is upregulated in metastatic cancers, a phenomenon known as the Warburg effect. Measurement of intracellular levels of 2PG is a tool for analyzing the glycolytic pathway in cancer cells.

Suitability

Suitable for the detection of 2-Phosphoglycerate in tissue and cell culture samples.

Principle

The 2-Phosphoglycerate Colorimetric/Fluorometric Assay Kit is a simple assay that measures 2PG in various samples (ranging from 2-10 nmole/well for the colorimetric assay and from 50-250 pmole/well for the fluorometric assay). 2PG concentration is determined by converting the 2PG to PEP and then to pyruvate. The pyruvate is oxidized, resulting in a colorimetric (570 nm) or fluorometric (λex = 535/ λem = 587 nm) product proportional to the amount of 2PG present.

Other Notes

The fluorometric assay is 10-100 times more sensitive than the colorimetric assay.

Kit Components Only

Product No.
Description

  • 2PG Assay Buffer

  • 2PG Probe, in DMSO

  • 2PG Enzyme Mix

  • 2PG Converter

  • 2PG Developer

  • 2PG Standard

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service