Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MAK081

Sigma-Aldrich

ADP Colorimetric Assay Kit II

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.84

usage

sufficient for 100 colorimetric tests

detection method

colorimetric

relevant disease(s)

hematological disorder; cancer

storage temp.

−20°C

General description

Adenosine diphosphate (ADP) is a nucleoside diphosphate that plays a critical role in energy transfer reactions.[1] ADP is produced from adenosine triphosphate via the action of ATPases. ADP also plays a critical role in platelet function.[2] ADP, stored in platelet-dense granules, is released upon platelet activation where it acts on purinergic receptors to mediate intracellular signaling and platelet aggregation.[3][4]

Application

ADP Colorimetric Assay Kit II has been used to measure the ADP level in infusion medium and variety of samples, particularly those containing reducing agents that may interfere with oxidase based assays.[5][6]

Suitability

Suitable for measuring ADP levels in samples that contain reducing substances, which may interfere with oxidase-based assays. Samples include tissue (liver, muscle, heart etc.) and cell culture (adherent or suspension cells).

Principle

The ADP Assay kit provides a simple and direct procedure for measuring ADP in a variety of samples. ADP concentration is determined by a coupled enzyme assay, which results in a colorimetric (450 nm) product proportional to the ADP present.

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of novel Akt1 inhibitors as anticancer agents using virtual co-crystallized pharmacophore generation.
Al-Sha?er M A, et al.
Journal of Molecular Graphics & Modelling, 62, 213-225 (2015)
Tomas A Schiffer et al.
American journal of physiology. Renal physiology, 315(3), F677-F681 (2018-05-31)
The kidneys receive ~25% of cardiac output, which is a prerequisite to maintain sufficient glomerular filtration rate. However, both intrarenal regional renal blood flow and tissue oxygen levels are heterogeneous with decreasing levels in the inner part of the medulla.
Liyuan Zhu et al.
Nature communications, 10(1), 1888-1888 (2019-04-25)
The use of nonsteroidal anti-inflammatory drugs that inhibit cyclooxygenase (COX)-1 and COX-2, increases heart failure risk. It is unknown whether microsomal (m) prostaglandin (PG) E synthase (S)-1, a target downstream of COX, regulates myocardial (M) ischemia/reperfusion (I/R) injury, a key
Katherine D.
Chemistry (Weinheim An Der Bergstrasse, Germany) (2016)
Blockade of adenosine diphosphate receptors P2Y(12) and P2Y(1) is required to inhibit platelet aggregation in whole blood under flow.
Nancy A, et al.
Blood, 98(12), 3340-3345 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service