Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MAK024

Sigma-Aldrich

Cobalt Assay

sufficient for 100 colorimetric tests

Synonym(s):

Cobalt Quantitation Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.84
Product MAK024 is not currently sold in your country. Contact Technical Service

usage

sufficient for 100 colorimetric tests

application(s)

cosmetics
food and beverages

detection method

colorimetric

storage temp.

room temp

General description

Cobalt is a transition metal that serves as a trace dietary mineral for all multicellular organisms. Cobalt is an important cofactor for the Vitamin B12 class of compounds where it occupies the center of the vitamin B12 corrin ring.[1] Cobalt can also be coordinated in the active site of the non-corrin containing metalloenzyme methionine aminopeptidase.[1]

Suitability

Suitable for quantitating cobalt concentrations in a variety of samples

Principle

The Cobalt Assay kit provides a simple and direct procedure for measuring cobalt in a variety of samples. In this assay, cobalt reacts with 2-mercaptoethanol under basic conditions to form a complex with a strong absorbance at 475 nm. Interference from the metal ions Fe2+, Cu2+, Ni2+, Zn2+, and Mn2+ is <10% at this wavelength. This assay gives a linear range of 10-50 nmoles of cobalt.

signalword

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

target_organs

Liver,Heart

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cobalt proteins.
Kobayashi M and Sakayu S
FEBS Journal, 261(1), 1-9 (1999)
Dong Gil Lee et al.
Cell death & disease, 11(3), 204-204 (2020-03-25)
Iron is an essential element for cellular functions, including those of neuronal cells. However, an imbalance of iron homeostasis, such as iron overload, has been observed in several neurodegenerative diseases, including Alzheimer's disease and Parkinson's disease. Iron overload causes neuronal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service