Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M9817

Sigma-Aldrich

Anti-MBD4 antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~65 kDa

species reactivity

mouse

concentration

~1 mg/mL

technique(s)

microarray: suitable
western blot: 0.5-1 μg/mL using nuclear extracts of K-562 cell line

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MBD4(8930)
mouse ... Mbd4(17193)

General description

Human MBD4 protein encodes an alternative spliced form which lacks C-terminal 42 amino acids.
Methyl-CpG binding domain protein 4 (MBD4) is a thymine glycosylase which possesses a carboxy-terminal DNA glycosylase domain and an amino-terminal MBD domain. It belongs to the methyl-CpG-binding protein family.

Immunogen

synthetic peptide corresponding to amino acids 541-554 of mouse MBD4 conjugated to KLH via an N-terminal added cysteine residue. The sequence is conserved in human.

Application

Anti-MBD4 antibody produced in rabbit has been used in immunoblotting and chromatin-immunoprecipitation (ChIP).

Biochem/physiol Actions

Methyl-CpG binding domain protein 4 (MBD4) plays an important role in DNA demethylation. It can remove mismatched bases which are present with a guanine. The preferred substrate for MBD4 is thymine over N4-ethenocytosine.
The methyl-CpG binding domain of MBD4 binds preferentially to 5-methylcytosine CpG-TpG mismatches, the primary product of deamination at methyl-CpG. The combined specificities of binding and catalysis indicates that this protein may function to minimize mutation at methylCpG. in vivo, the DNA repair properties of MBD4 suggests that it plays a role in suppressing mutability and tumorigenesis.

Physical form

Solution in 0.01 M phosphate buffered saline containing 1% bovine serum albumin (BSA) and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MBD4 cooperates with DNMT1 to mediate methyl-DNA repression and protects mammalian cells from oxidative stress
Laget S, et al.
Epigenetics, 9(4), 546-556 (2014)
The thymine glycosylase MBD4 can bind to the product of deamination at methylated CpG sites
Hendrich B, et al.
Nature, 401(6750), 301-301 (1999)
Identification and characterization of a family of mammalian methyl CpG-binding proteins
Hendrich B and Bird A
Genetics research, 72(1), 59-72 (1998)
Sophie Laget et al.
Epigenetics, 9(4), 546-556 (2014-01-18)
Oxidative stress induces genome-wide remodeling of the chromatin structure. In this study, we identify Methyl-CpG Binding Protein 4 (MBD4), a multifunctional enzyme involved in DNA demethylation, base excision repair, and gene expression regulation, as an essential factor in response to
Enhanced CpG mutability and tumorigenesis in MBD4-deficient mice
Millar CB, et al.
Science, 297(5580), 403-405 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service