Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M8379

Sigma-Aldrich

7-Methyl-DL-tryptophan

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N2O2
CAS Number:
Molecular Weight:
218.25
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

7-Methyl-DL-tryptophan, crystalline

assay

≥98% (HPLC)

form

crystalline

color

off-white

storage temp.

2-8°C

SMILES string

Cc1cccc2c(CC(N)C(O)=O)c[nH]c12

InChI

1S/C12H14N2O2/c1-7-3-2-4-9-8(6-14-11(7)9)5-10(13)12(15)16/h2-4,6,10,14H,5,13H2,1H3,(H,15,16)

InChI key

KBOZNJNHBBROHM-UHFFFAOYSA-N

Biochem/physiol Actions

7-Methyl-DL-tryptophan is a tryptophan analog.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Barone et al.
Plant cell reports, 27(3), 509-517 (2007-12-07)
Effective selectable markers are needed for basic research and commercial applications that do not involve antibiotic or herbicide resistance. A novel selection system based on a feedback-insensitive anthranilate synthase alpha-subunit of tobacco (ASA2) as selectable marker using either 4-methylindole (4MI)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service