Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M7931

Sigma-Aldrich

Monoclonal Anti-Mcm7 antibody produced in mouse

clone DCS-141, ascites fluid

Synonym(s):

Anti-Cdc47

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

DCS-141, monoclonal

mol wt

antigen ~80 kDa

contains

15 mM sodium azide

species reactivity

human, Xenopus

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: 1:2,000 using HeLa cell nuclear preparation

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MCM7(4176)

General description

Minichromosome maintenance complex component 7 (MCM7) is part of the pre-replication complex.
Monoclonal Anti-MCM7 (CDC47) (mouse IgG1 isotype) is derived from the DCS-141 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice. The MCM7 gene is mapped to human chromosome 7q22.1.

Specificity

Recognizes human Mcm7 and also a weaker lower molecular weight doublet.

Immunogen

recombinant human Mcm7 (Cdc47) protein

Application

Monoclonal Anti-Mcm7 antibody produced in mouse has been used in:
  • immunoblotting
  • immunocytochemistry
  • immunoprecipitation

Biochem/physiol Actions

MCM7 levels do not vary with cell cycle, but the expression and nascent synthesis of MCM7 occurs late in the cell cycle, coinciding with mitosis. MCM7 immunolocalization may be used as an index of cell proliferation in tissue sections. Monoclonal antibody reacting specifically with MCM7 is also a useful tool in the investigation of the molecular mechanisms behind DNA replication and its restriction to S phase. MCM7 may play a role in normal and neoplastic cell growth. MCM7 favors hepatocellular carcinoma progression.
Minichromosome maintenance complex component 7 (MCM7) has been shown to possess DNA helicase activity and is crucial for initiating replication.

Target description

Mcm7 (Cdc47) belongs to the minichromosome maintenance family.

Physical form

The product is provided as ascites fluid with 15 mM sodium azide (see MSDS) as a preservative.

Storage and Stability

For continuous use, store at 2-8 degree C for up to one month. For extended storage freeze in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takatsugu Kan et al.
Gastroenterology, 136(5), 1689-1700 (2009-05-08)
Barrett's esophagus (BE) is a highly premalignant disease that predisposes to the development of esophageal adenocarcinoma (EAC); however, the involvement of microRNAs (miRs) in BE-EAC carcinogenic progression is not known. Esophageal cultured cells (HEEpiC, QhTRT, ChTRT, GihTRT, and OE-33) and
B K Tye
Annual review of biochemistry, 68, 649-686 (2000-06-29)
The MCM proteins are essential replication initiation factors originally identified as proteins required for minichromosome maintenance in Saccharomyces cerevisiae. The best known among them are a family of six structurally related proteins, MCM2-7, which are evolutionally conserved in all eukaryotes.
Katerina Vrzalikova et al.
Pathogens (Basel, Switzerland), 1(2), 83-101 (2012-01-01)
B-lymphocyte-induced maturation protein 1 (BLIMP1) exists as two major isoforms, α and β, which arise from alternate promoters. Inactivation of the full length BLIMP1α isoform is thought to contribute to B cell lymphomagenesis by blocking post-germinal centre (GC) B cell
Masatoshi Takagi et al.
The FEBS journal, 275(24), 6109-6122 (2008-11-20)
Sister chromatid cohesion is important for the correct alignment and segregation of chromosomes during cell division. Although the cohesin complex has been shown to play a physical role in holding sister chromatids together, its loading onto chromatin is not sufficient
Kai Qu et al.
Cell death & disease, 8(2), e2603-e2603 (2017-02-10)
DNA replication is a central procedure of cell proliferation, whereas aberrant DNA replication is indicated to be a driving force of oncogenesis. Minichromosome maintenance complex component 7 (MCM7) plays an essential role in initiating DNA replication. To investigate the potential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service