One unit will hydrolyze 1 μmole of p-nitrophenyl β-D-mannopyranoside to p-nitrophenol (measured at 400 nm) and D-mannose per minute at pH 4.0 at 37 °C.
Physical form
Enzyme is lyophilized from 10 mM sodium acetate buffer, pH 4.0, containing BSA and sodium chloride.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
Applied biochemistry and biotechnology, 167(1), 73-80 (2012-04-25)
A novel biosensor strain (Escherichia coli ALM403) that responded to N-acyl homoserine lactone (AHL) was constructed using a luxR-Plux cassette as a regulatory sequence and β-mannanase as a reporter gene. Dinitrosalicylic acid method was used to detect the response of
Statistical modeling was applied for describing structural features of β-(1→4)-D-galactomannans. According to the model suggested theoretical ratios of limiting degrees of locust bean, tara gum and guar gum galactomannan conversions by two β-(1→4)-mannanases of different origin (Myceliophthora thermophila and Trichoderma
Journal of applied microbiology, 113(4), 798-806 (2012-07-14)
To screen and select the Bacillus spp. from Tua-nao of northern Thailand for fermented corticate soybean meal (FCSBM) production. After isolation of Bacillus spp. from Tua-nao was carried out, cellulase, hemicellulases (i.e., β-mannanase and xylanase) and phytase production by isolated
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology, 162(1-3), 24-33 (2012-03-01)
A β-D-mannosidase (EC 3.2.1.25) with a molecular mass of approximately 100 kDa was purified from the digestive fluid of a marine gastropod Aplysia kurodai by ammonium sulfate fractionation followed by column chromatographies on TOYOPEARL Butyl-650 M, TOYOPEARL DEAE-650 M, and
Journal of plant physiology, 169(12), 1125-1133 (2012-06-05)
Cell walls of tomato fruit contain hemicellulosic mannans that may fulfill a structural role. Two populations were purified from cell walls of red ripe tomato tissue and named galactoglucomannan-glucuronoxylan I and II (GGM-GX I and II), respectively. Both polysaccharides not
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.