M3640
Mannan from Saccharomyces cerevisiae
prepared by detergent extraction
Synonym(s):
D-Mannan
About This Item
Recommended Products
biological source
Saccharomyces cerevisiae
form
powder
optical activity
[α]25/D 73 to 82 °, c = 1% (w/v) in water
impurities
≤5% ethanol
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Application
Preparation Note
Other Notes
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service