Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L9913

SAFC

LYN A human

≥70% (SDS-PAGE), recombinant, expressed in baculovirus infected insect cells, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in baculovirus infected insect cells

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

≥600 nmol/min·mg

mol wt

53-56 kDa

UniProt accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... LYN(4067)

Biochem/physiol Actions

Catalyzes the phosphorylation of the terminal tyrosine residues of the Tec protein, a downstream protein in intracellular signaling pathways.

Other Notes

Histidine tagged.

Unit Definition

Using 50 mM HEPES, pH 7.5, 10 mM MgCl2, 2.5 mM DTT, 10% glycerol, 0.01% Triton X-100, 200 μM ATP, 200 μg/mL poly [Glu, Tyr] 4:1 and trace [32P]-gamma ATP at 30°C

Physical form

Solution in 50 mM Tris, pH 7.5, 0.05 mM EDTA, 1 mM DTT, 100 mM NaCl, 0.05% NP-40, and 50% glycerol.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service