Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L6890

Sigma-Aldrich

Lectin from Sambucus nigra (elder)

lyophilized powder

Synonym(s):

Sambucus nigra agglutinin, SNA

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202

form

lyophilized powder

Quality Level

potency

≤4 μg/mL agglutination activity

composition

Protein, 20-40% Lowry

storage temp.

−20°C

Biochem/physiol Actions

SNA is isolated from the inner bark (bast tissue) of elder stems and branches. The lectin is not blood group specific but has an affinity for α-NeuNAc-[2→6]-Gal, α-NeuNAc-[2→6]-GalNAc, and, to a lesser extent, α-NeuNAc-[2→3]-Gal residues.

Packaging

Package size based on protein content

Physical form

Lyophilized powder containing NaCl

Preparation Note

Affinity purified by a modification of Broekaert.[1]

Analysis Note

Agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 7.3, of a 1 mg/ml solution. The activity is the lowest concentration to agglutinate a 2% suspension of human blood group A erythrocytes after 1 hr incubation at 25 °C.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W F Broekaert et al.
The Biochemical journal, 221(1), 163-169 (1984-07-01)
A lectin was isolated from elder (Sambucus nigra) bark by affinity chromatography on fetuin-agarose. It is a tetrameric molecule (Mr 140000) composed of two different subunits of Mr 34500 and 37500 respectively, held together by intramolecular disulphide bridges. The lectin
H J Hoffmann et al.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology, 33(7), 930-935 (2003-07-16)
Atopy is closely associated with the cellular T helper type-2 (Th2) phenotype, that is dominated by the pleiotrophic cytokine IL-4. The cellular source of IL-4 has yet to be determined, although basophils have been proposed. Eosinophils and mast cells are

Articles

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service