British medical bulletin, 56(3), 761-772 (2001-03-20)
Gonadotropin-releasing hormone (GnRH) is a decapeptide (pGlu-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-Gly.NH2) which is produced from a precursor polypeptide in hypothalamic neurons and secreted in a pulsatile fashion to stimulate the secretion of LH and FSH via its interaction with a cognate receptor on gonadotropes.
Biochemistry and cell biology = Biochimie et biologie cellulaire, 64(12), 1372-1377 (1986-12-01)
Immunochemical studies on monoclonal and conventional anti-LHRH antibodies have been reported. The association constants (Ka) were in the order of 10(9)-10(10) L/mol when calculated from Scatchard and Steward-Petty plots. Heterogeneity indices (a) calculated from Sips plot were nearly 1.0, indicating
The correlation between neuronal mechanism of anxiety and neuroanatomic expression/neuromodulatory role of gonadotropin-releasing hormone (GnRH), points to a role of GnRH in the modulation of anxiety. Therefore, we investigated the influence of GnRH agonists and antagonist on the anxiety-like behavior
Questions
Reviews
★★★★★ No rating value