Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

K1513

Sigma-Aldrich

Kendomycin from Streptomyces violaceoruber

solid

Synonym(s):

(−)-TAN 2162

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H42O6
CAS Number:
Molecular Weight:
486.64
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
neoplastics

mode of action

protein synthesis | interferes

storage temp.

−20°C

SMILES string

CC(C)CC(N)C(=O)NC(CCCNC(N)=N)C(=O)NC(CCCNC(N)=N)C(=O)NC(C)C(=O)NC(CO)C(=O)NC(C(C)C)C(=O)NC(C)C(O)=O

InChI

1S/C32H61N13O9/c1-15(2)13-19(33)25(48)42-21(10-8-12-39-32(36)37)27(50)43-20(9-7-11-38-31(34)35)26(49)40-17(5)24(47)44-22(14-46)28(51)45-23(16(3)4)29(52)41-18(6)30(53)54/h15-23,46H,7-14,33H2,1-6H3,(H,40,49)(H,41,52)(H,42,48)(H,43,50)(H,44,47)(H,45,51)(H,53,54)(H4,34,35,38)(H4,36,37,39)

InChI key

PPBZOEOSJVJLRX-UHFFFAOYSA-N

General description

Chemical structure: peptide

Biochem/physiol Actions

Potent endothelin receptor antagonist. Antiosteoporotic compound. Shows also remarkable antibacterial and cytotoxic activity. A putative cellular target for cytotoxicity has been identified: inhibition or altered regulation of the proteasome.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure and biosynthesis of kendomycin, a carboxylic ansa-compound from Streptomyces. .
Zeeck, A. , and Bode, H.B.
Journal of the Chemical Society. Perkin Transactions 1, 323-323 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service