Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I3653

Sigma-Aldrich

Anti-Interferon β antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-IFN-β

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

western blot: 0.1-2.0 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IFNB1(3456)

General description

Type I interferons are proteins produced in response to inducers like viral infection and can be divided into α and β subtypes. IFN-β provokes high antiproliferation response in certain cells like embryonal carcinoma, melanoma and melanocytes. It also has high potency in treatment of multiple sclerosis and certain cancers. Anti-interferon β antibody can be used in western blot. Goat anti-interferon β antibody reacts specifically with human interferon-β and has shown no cross reactivity with recombinant human interferon-γ.

Specificity

The antibody will neutralize the biological activity of recombinant human IFN-β. It will not block the biological activity of recombinant human IFN-α or IFN-γ.

Immunogen

recombinant human IFN-β, expressed in Escherichia coli.

Application

Anti-interferon β antibody can be used in immunoblotting. It can also be used to neutralize the bioactivity of recombinant human interferon-β.

Physical form

Lyophilized from a solution in phosphate buffered saline containing carbohydrates.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Belhumeur et al.
Molecular and cellular biology, 13(5), 2846-2857 (1993-05-01)
In the current study, we have addressed the role of interferons (IFNs) in controlling the differentiation of pluripotent P19 embryonal carcinoma (EC) cells. Blocking IFN activity in the culture medium of differentiating cells with antibodies leads to a strong decrease

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service