I2158
Anti-phospho-Insulin Receptor/Insulin-Like Growth Factor-1 Receptor (pTyr1162/1163) antibody produced in rabbit
affinity isolated antibody, buffered aqueous glycerol solution
About This Item
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous glycerol solution
species reactivity
human, rat
technique(s)
immunocytochemistry: suitable
western blot: 1:1000 using CHO T-cells transfected with a vector containing insulin receptor and stimulated with insulin.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... IGF1R(3480)
rat ... Igf1r(25718)
General description
Anti-phospho-IR/IGF1R (pTyr1162/1163) specifically recognizes human and rat insulin receptor and insulin-like growth factor-1 receptor phosphorylated at tyrosines 1162 and 1163.
Immunogen
Application
Target description
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service