Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I2033

Sigma-Aldrich

Anti-phospho-Insulin Receptor/Insulin-Like Growth Factor-1 Receptor (pTyr1158/1162/1163) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-IR/IGF1R

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 0.1-1.0 mg/mL using stimulated CHO-T cells

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IGF1R(3480)

General description

Insulin-like growth factors -1 and -2 are important regulators of signalling that mediates growth and metabolism in cells. Insulin-like growth factor -1 shares structural homology with proinsulin and is mainly produced by liver. IGF-1 acts via specific receptor, IGF-1R, a heterodimeric tyrosine kinase receptor that signals to various second messenger pathways such as MAPK, Raf/Ras and PI3K cascades. IGF 1R is fully activated by phosphorylation of tyrosine residues at Tyr1158, Tyr1162 and Tyr1163. The effect of IGF-1 is by endocrine, paracrine and autocrine signalling. The effects of IGF-1R signalling are important at many developmental levels from cells to organ to organism, growth, differentiation, anti-apoptosis, proliferation and transformation. Irregular IGF-1 signaling has been implicated in many cancers including melanoma, breast, lung, colon and prostate
Anti-phospho-IR/IGF1R (pTyr1158/1162/1163) specifically recognizes human receptor and insulin-like growth factor-1 receptor phosphorylated at tyrosines 1158, 1162 and 1163. It may cross react with mouse and rat IR/IGF1R (100% homology).

Immunogen

synthetic phosphopeptide derived from the region of IR that contains tyrosines 1158, 1162, and 1163.

Application

Anti-phospho-IR/IGF1R (pTyr1158/1162/1163) antibody may be used for immunoblotting at a working concentration of 0.1-1.0 μg/ml using whole cell lysates from CHO-T cells.

Physical form

Solution in Dulbecco′s phosphate buffered saline, pH 7.3, containing 50% glycerol, 1 mg/mL BSA (IgG and protease-free) and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lysophosphatidylcholine inhibits insulin-induced akt activation through protein kinase C-α in vascular smooth muscle cells
Motley ED et al
Hypertension, 3, 508-512 (2002)
Rosaria Ottanà et al.
Molecules (Basel, Switzerland), 26(2) (2021-01-14)
Diabetes mellitus (DM) is a complex disease which currently affects more than 460 million people and is one of the leading cause of death worldwide. Its development implies numerous metabolic dysfunctions and the onset of hyperglycaemia-induced chronic complications. Multiple ligands
Rosanna Maccari et al.
European journal of medicinal chemistry, 252, 115270-115270 (2023-03-20)
Type 2 diabetes mellitus (T2DM) is a serious chronic disease with an alarmingly growing worldwide prevalence. Current treatment of T2DM mainly relies on drug combinations in order to control blood glucose levels and consequently prevent the onset of hyperglycaemia-related complications.
H Yu et al.
Journal of the National Cancer Institute, 91(2), 151-156 (1999-01-29)
Insulin-like growth factors (IGFs), in particular IGF-I and IGF-II, strongly stimulate the proliferation of a variety of cancer cells, including those from lung cancer. To examine the possible causal role of IGFs in lung cancer development, we compared plasma levels
J M Chan et al.
Science (New York, N.Y.), 279(5350), 563-566 (1998-02-07)
Insulin-like growth factor-I (IGF-I) is a mitogen for prostate epithelial cells. To investigate associations between plasma IGF levels and prostate cancer risk, a nested case-control study within the Physicians' Health Study was conducted on prospectively collected plasma from 152 cases

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service