Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA030088

Sigma-Aldrich

Anti-LMX1A antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2

Synonym(s):

Anti-LIM homeobox transcription factor 1, α, Anti-LMX1, Anti-LMX1.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL

immunogen sequence

QRAKMKKLARRQQQQQQDQQNTQRLSSAQTNGGGSAGMEGIMNPYTALPTPQQLLAIEQSVYSSDPFRQGLT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... LMX1A(4009)

General description

The gene LMX1A (LIM homeobox transcription factor 1 α) is mapped to human chromosome 1q23.3. It is a paralogue of Lhx (LIM homeobox family) proteins.

Immunogen

LIM homeobox transcription factor 1, α recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

LMX1A (LIM homeobox transcription factor 1 α) is a transcription factor which is required for the development of dopaminergic neurons. It plays an important role in the dopaminergic pathway and is associated with schizophrenia and altered cognitive functioning. Mutation in the gene is also associated with risk of Parkinson′s disease. LMX1A gene is upregulated in advanced stages of pancreatic ductal adenocarcinoma. It is hypermethylated in squamous cell carcinoma and is a suppressor of metastasis in cervical cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77007.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dopaminergic control of autophagic-lysosomal function implicates Lmx1b in Parkinson's disease.
Laguna A, et al.
Nature Neuroscience, 18, 826-835 (2015)
High-resolution genomic copy number profiling of primary intraocular lymphoma by single nucleotide polymorphism microarrays.
Wang L, et al.
Cancer Science, 105, 592-599 (2014)
Preliminary evidence that allelic variation in the LMX1A gene influences training-related working memory improvement.
Bellander M, et al.
Neuropsychologia, 49, 1938-1942 (2011)
Francesca Sgualdino et al.
Methods in molecular biology (Clifton, N.J.), 2764, 225-245 (2024-02-23)
Organoids are a powerful model system to explore the role of mechanical forces in sculpting emergent tissue cytoarchitecture. The modulation of the mechanical microenvironment is most readily performed using synthetic extracellular matrices (ECM); however, such materials provide passive, rather than
The role of Sp1 and EZH2 in the regulation of LMX1A in cervical cancer cells.
Lin WC, et al.
Biochimica et Biophysica Acta, 1833, 3206-3201 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service