Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA019614

Sigma-Aldrich

Anti-AHRR antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-Apoptosis-linked gene 2 protein, Anti-PDCD6, Anti-Probable calcium-binding protein ALG-2, Anti-Programmed cell death protein 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

immunogen sequence

RDVGEDQVHPPLCHFPQRSLQHQLPQPGAQRFATRGYPMEDMKLQGVPMPPGDLCGPTLLLDVSIKMEKDSGCEGAADGCVPSQVWLGASDRSHPATF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDCD6(57491)

General description

AHRR (Aryl-hydrocarbon receptor repressor) is a aryl hydrocarbon receptor (AHR) target gene belonging to the bHLH-PAS protein.[1] It is composed of a 2094-bp mRNA encoded by ten exons. It is localized to chromosome 5p. It possesses structural similarity with AHR (Aryl hydrocarbon receptor) and ARNT (AHR nuclear translocator).

Immunogen

Programmed cell death protein 6 recombinant protein epitope signature tag (PrEST)

Application

Anti-AHRR antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues.[1] These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

AHRR (Aryl-hydrocarbon receptor repressor) plays a crucial role in the regulation of cell growth and differentiation. It is involved in the ligand-induced repression activities of AhR-regulated genes by sequestering ARNT. AHRR also play an important role in TCDD-stimulated AhR signaling pathway. It negatively controls AHR, which plays an essential role in mediating the toxic and biochemical effects of 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74244.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sunny Y Yang et al.
Archives of toxicology, 92(1), 225-240 (2017-07-07)
The aryl hydrocarbon receptor (AHR) mediates the toxic actions of environmental contaminants, such as 2,3,7,8-tetrachlorodibenzo-ρ-dioxin (TCDD), and also plays roles in vascular development, the immune response, and cell cycle regulation. The AHR repressor (AHRR) is an AHR-regulated gene and a
Damian Brauze et al.
Toxicology letters, 167(3), 212-220 (2006-10-31)
The aryl hydrocarbon receptor (AhR) mediates a variety of biological responses to ubiquitous environmental pollutants. AhR together with ARNT, AhRR, HIF1alpha represent a novel basic helix-loop-helix/PAS family of transcriptional regulators. Their interplay may affect the xenobiotic response. In this study
Thorsten Bernshausen et al.
Archives of toxicology, 80(4), 206-211 (2005-10-06)
The Aryl hydrocarbon receptor repressor (AhRR) is a new member of bHLH-PAS proteins which is important in the regulation of cell growth and differentiation. The AhRR shares structural similarities with Aryl hydrocarbon receptor (AhR) and AhR nuclear translocator (ARNT). The
T Watanabe et al.
Journal of human genetics, 46(6), 342-346 (2001-06-08)
The diversity of biological effects resulting from exposure to dioxin may reflect the ability of this environmental pollutant to alter gene expression by binding to the arylhydrocarbon receptor (AHR) gene and related genes. AHR function may be regulated by structural
Laura MacPherson et al.
International journal of molecular sciences, 15(5), 7939-7957 (2014-05-09)
The aryl hydrocarbon receptor (AHR) regulates the toxic effects of 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD). The AHR repressor (AHRR) is an AHR target gene and functions as a ligand-induced repressor of AHR; however, its mechanism of inhibition is controversial. Recently, we reported that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service