Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA012383

Sigma-Aldrich

Anti-SLC29A1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Equilibrative NBMPR-sensitive nucleoside transporter, Anti-Equilibrative nitrobenzylmercaptopurine riboside-sensitive nucleoside transporter, Anti-Equilibrative nucleoside transporter 1, Anti-Nucleoside transporter, es-type, Anti-Solute carrier family 29 member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 1:100- 1:250
immunofluorescence: 1-4 μg/mL
immunohistochemistry: 1:20- 1:50

immunogen sequence

TQYFTNRLDMSQNVSLVTAELSKDAQASAAPAAPLPERNSL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC29A1(2030)

Immunogen

Equilibrative nucleoside transporter 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SLC29A1 (Solute carrier family 29, member 1) encodes an equilibrative nucleoside transporter 1 (hENT1) protein in humans. It is expressed in human hepatocytes, which indicates an essential role in controlling hENT1 expression levels in those cells. It mainly performs in the nucleotide (purines and pyrimidines) transport into the tumor cells. It has been reported that SLC29A1 plays a vital role in pancreatic cancer as a predictive biomarker for identifying gemcitabine efficacy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72507

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Judith E K Henning et al.
The Journal of clinical endocrinology and metabolism, 102(11), 4323-4332 (2017-11-02)
Adrenocortical carcinoma (ACC) is rare and confers an unfavorable prognosis in advanced stages. Other than combination chemotherapy with cisplatin, etoposide, doxorubicin, and mitotane, the second- and third-line regimens are not well-established. Gemcitabine (GEM)-based chemotherapy was suggested in a phase 2
Beatrice Mohelnikova-Duchonova et al.
Pancreatology : official journal of the International Association of Pancreatology (IAP) ... [et al.], 13(6), 558-563 (2013-11-28)
Although systemic chemotherapy significantly improves the overall survival of pancreatic cancer patients, the prognosis remains extremely poor. The development of a drug resistance, either de novo or induced resistance, significantly limits the effectiveness of chemotherapy. SLC29A1 gene encodes human equilibrative
Steffen Ormanns et al.
European journal of cancer (Oxford, England : 1990), 50(11), 1891-1899 (2014-05-27)
The role of human equilibrative nucleoside transporter 1 (hENT1) as a predictive biomarker for gemcitabine efficacy in advanced pancreatic cancer remains unclear to date. AIO-PK0104 was a German multicenter phase III trial comparing gemcitabine/erlotinib followed by capecitabine (GEC) with capecitabine/erlotinib
Tomomi Furihata et al.
Drug metabolism and pharmacokinetics, 29(4), 325-332 (2014-02-14)
Human equilibrative nucleoside transporter 1 (hENT1) transports various nucleoside analogues into cells. Although the single hENT1 promoter region (P1) and the mRNA isoform (a1) have been characterized previously, we have recently identified additional promoter regions P2 and P3 (which primarily

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service