Merck
  • Human equilibrative nucleoside transporter 1 is not predictive for gemcitabine efficacy in advanced pancreatic cancer: translational results from the AIO-PK0104 phase III study with the clone SP120 rabbit antibody.
Human equilibrative nucleoside transporter 1 is not predictive for gemcitabine efficacy in advanced pancreatic cancer: translational results from the AIO-PK0104 phase III study with the clone SP120 rabbit antibody.
European journal of cancer (Oxford, England : 1990) (2014-05-27)
Steffen Ormanns, Volker Heinemann, Mitch Raponi, Jeff Isaacson, Rüdiger P Laubender, Michael Haas, Stephan Kruger, Axel Kleespies, Elaina Mann, Mike Bartosiewicz, Thomas Kirchner, Stefan Boeck
ABSTRACT
The role of human equilibrative nucleoside transporter 1 (hENT1) as a predictive biomarker for gemcitabine efficacy in advanced pancreatic cancer remains unclear to date. AIO-PK0104 was a German multicenter phase III trial comparing gemcitabine/erlotinib followed by capecitabine (GEC) with capecitabine/erlotinib followed by gemcitabine (CEG) in advanced pancreatic cancer. Archival tumour tissue from 169 of the 274 eligible study patients was available for a central and standardised immunohistochemistry staining for hENT1 expression using the SP120 rabbit monoclonal anti-hENT1 antibody. Within a retrospective translational subgroup analysis, biomarker data were correlated with efficacy end-points. Thirty-nine out of 130 fresh-cut slides were scored as hENT1(high) (30%), whereas 91 samples were classified as hENT1(low) (70%). For the 62 patients randomised to CEG median overall survival was estimated with 6.4 months in the hENT1(low) compared to 6.9 months in the hENT1(high) subgroup (Hazard Ratio (HR) 0.88, 95% confidence interval (CI) 0.48-1.61, p=0.67). For the 68 patients randomised to GEC survival was 5.7 months in the hENT1(low) compared to 4.4 months in the hENT1(high) subgroup (HR 1.16, 95% CI 0.69-1.96, p=0.57). In 101 patients receiving gemcitabine at any time during study treatment (either within the 1st- or 2nd-line setting) hENT1(low) cases had a median overall survival of 7.5 months and hENT1(high) patients an overall survival of 4.4 months (HR 1.30, 95% CI 0.84-2.03, p=0.24), respectively. Within this subgroup analysis from Arbeitsgemeinschaft Internistische Onkologie-pancreatic cancer (AIO-PK0104), no evidence supporting the use of hENT1 as a predictive biomarker for gemcitabine efficacy in patients with advanced pancreatic cancer was found.