Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA012065

Sigma-Aldrich

Anti-TNK1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD38 negative kinase 1, Anti-Non-receptor tyrosine-protein kinase TNK1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

PSEACCVRDVTEPGALRMETGDPITVIEGSPDSTIWKGQNGRTFKVGSFPASAVTLADAGGLPATRPVHRGTPARGDQHPGSIDGDRKKANLWDAPPARGQRRNMPLERMKGISRSLESVLSLG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNK1(8711)

General description

TNK1 (Tyrosine kinase, non-receptor, 1) is a novel 72kDa nonreceptor tyrosine kinase protein beloniging to the ACK (Activated CDC42 kinase) family of non-receptor tyrosine kinases. It consists of an N-terminal kinase domain, a putative Src Homology 3 (SH3) domain at the C-terminal end, and a proline-rich tail. TNK1 is expressed in all fetal tissues such as cord blood, bone marrow and adult tissues such as prostate, testis, ovary, small intestine and colon. In addition to normal tissues, its expression also have been found in the several leukemia cell lines.

Immunogen

Non-receptor tyrosine-protein kinase TNK1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

TNK1 (Tyrosine kinase, non-receptor, 1) is majorly involved in the phospholipid signaling pathways. In addition to the kinase activity, it also negetively regulates Ras-Raf1-MAPK pathway. Studies have been suggested that TNK1 may possess tumor suppressor like properties as well as capacity of tumor necrosis factor-α-induced apoptosis. It may participate in various signaling pathways during fetal development and growth.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71457

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D M Felschow et al.
Biochemical and biophysical research communications, 273(1), 294-301 (2000-06-30)
Tnk1 is a nonreceptor tyrosine kinase cloned from CD34+/Lin-/CD38- hematopoietic stem/progenitor cells. The cDNA predicts a 72-kDa protein containing an NH(2)-terminal kinase, a Src Homology 3 (SH3) domain, and a proline-rich (PR) tail. We generated rabbit antiserum to a GST-Tnk1(SH3)
Identification of activated Tnk1 kinase in Hodgkin's lymphoma.
T-L Gu et al.
Leukemia, 24(4), 861-865 (2010-01-22)
G T Hoehn et al.
Oncogene, 12(4), 903-913 (1996-02-15)
Degenerate PCR was employed to identify novel tyrosine kinase genes from an enriched population of human umbilical cord blood hematopoietic stem/progenitor cells. One novel tyrosine kinase gene, designated Tnk1, was cloned. The sequence of the complete Tnk1 coding region predicts

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service