Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

HPA001216

Sigma-Aldrich

Anti-PIK3R1 antibody produced in rabbit

enhanced validation

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-PI3-kinase p85-subunit α antibody produced in rabbit, Anti-PI3K antibody produced in rabbit, Anti-Phosphatidylinositol 3-kinase regulatory subunit α antibody produced in rabbit, Anti-PtdIns-3-kinase p85-α antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$530.00

$530.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$530.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$530.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

LADAFKRYLLDLPNPVIPAAVYSEMISLAPEVQSSEEYIQLLKKLIRSPSIPHQYWLTLQYLLKHFFKLSQTSSKNLLNARVLSEIFSPMLFRFSAASSDNTENLIKVIEILISTEWNERQPAPALPPKPPKPTTVANNGMNNNMSL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PIK3R1(5295)

Looking for similar products? Visit Product Comparison Guide

Immunogen

Phosphatidylinositol 3-kinase regulatory subunit α recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-PIK3R1 antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

PIK3R1 (phosphoinositide-3-kinase, regulatory subunit 1) gene encodes the 85kDa regulatory subunit of phosphatidylinositol 3-kinase. The enzyme contains a 110kDa catalytic subunit that phosphorylates inositol ring of phosphatidylinositol at the 3′ position. It is essential for the insulin-stimulated increase in glucose uptake and glycogen synthesis in tissues that are insulin-sensitive. Mutation in this gene causes SHORT syndrome characterized by short stature, hyperextensibility of joints, ocular depression, Rieger anomaly and teething delay.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78872

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of tropomyosin-receptor-kinase B and phosphoinositide-3-kinase/protein kinase B signaling cascade
Yen, Tran Phi Hoang.
Indonesian Journal of Pharmacy, 25, 61-67 (2014)
Carlotta Costa et al.
Cancer cell, 26(4), 445-447 (2014-10-15)
A growing number of mutations in PIK3R1, the gene that encodes for the p85α regulatory subunit of PI3K, have been recently identified. In this issue of Cancer Cell, Cheung and colleagues describe two neomorphic PIK3R1 mutants prevalent in endometrial and
Clea Bárcena et al.
BMC medical genetics, 15, 51-51 (2014-06-03)
SHORT syndrome is a rare autosomal dominant condition whose name is the acronym of short stature, hyperextensibility of joints, ocular depression, Rieger anomaly and teething delay (MIM 269880). Additionally, the patients usually present a low birth weight and height, lipodystrophy
Diana Mandelker et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(40), 16996-17001 (2009-10-07)
Mutations in oncogenes often promote tumorigenesis by changing the conformation of the encoded proteins, thereby altering enzymatic activity. The PIK3CA oncogene, which encodes p110alpha, the catalytic subunit of phosphatidylinositol 3-kinase alpha (PI3Kalpha), is one of the two most frequently mutated
Franck Mauvais-Jarvis et al.
The Journal of clinical investigation, 109(1), 141-149 (2002-01-10)
A critical component of insulin action is the enzyme phosphoinositide (PI) 3-kinase. The major regulatory subunits of PI 3-kinase, p85alpha and its splice variants, are encoded by the Pik3r1 gene. Heterozygous disruption of Pik3r1 improves insulin signaling and glucose homeostasis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service