Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H8262

SAFC

Hanks′ Balanced Salt solution

With sodium bicarbonate, sterile-filtered

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

sterility

sterile-filtered

form

liquid

components

NaHCO3: yes

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hae-Ran Park et al.
BMC cancer, 9, 85-85 (2009-03-19)
Although cisplatin is one of the most effective chemotherapeutic agents, cisplatin alone does not achieve a satisfactory therapeutic outcome. Also cisplatin accumulation shows toxicity to normal tissues. In this study, we examined the possibility of HemoHIM both to enhance anticancer
Jamie L Fornek et al.
Journal of virology, 83(21), 11102-11115 (2009-08-21)
The transmission of H5N1 influenza viruses from birds to humans poses a significant public health threat. A substitution of glutamic acid for lysine at position 627 of the PB2 protein of H5N1 viruses has been identified as a virulence determinant.
Raju V S Rajala et al.
Journal of neurochemistry, 110(5), 1648-1660 (2009-07-07)
The insulin receptor (IR) and IR signaling proteins are widely distributed throughout the CNS. IR signaling provides a trophic signal for transformed retinal neurons in culture and we recently reported that deletion of IR in rod photoreceptors by Cre/lox system
Zheng-Shun Wen et al.
Marine drugs, 9(6), 1038-1055 (2011-07-13)
The study was conducted to investigate the promoted immune response to ovalbumin in mice by chitosan nanoparticles (CNP) and its toxicity. CNP did not cause any mortality or side effects when mice were administered subcutaneously twice with a dose of
Hideaki Oike et al.
PloS one, 6(8), e23709-e23709 (2011-09-09)
The circadian clock is closely associated with energy metabolism. The liver clock can rapidly adapt to a new feeding cycle within a few days, whereas the lung clock is gradually entrained over one week. However, the mechanism underlying tissue-specific clock

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service