Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H6783

Sigma-Aldrich

S-Hexylglutathione-Agarose

saline suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

form

saline suspension

extent of labeling

≥5 μmol per mL

matrix

Cross-linked 4% beaded agarose

matrix activation

epoxy

matrix attachment

amino

matrix spacer

12 atoms

storage temp.

2-8°C

Application

S-Hexylglutathione-agarose is an agarose conjugate in saline suspension used in affinity chromatography, protein chromatography, purification and detection, and recombinant protein expression and analysis. S-Hexylglutathione agarose has been used to inform vaccine development for major global diseases in livestock that infect humans, including the parasite Fasciola hepatica.

Physical form

Suspension in 0.5 M NaCl containing preservative

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preliminary Characterization of Glutathione <I>S</I>-Transferases That Accumulate in Callus Cells of Pumpkin (<I>Cucurbita maxima</I> Duch.).
Fujita, M., et al.
Plant & Cell Physiology, 35(2), 275-282 (1994)
Mohammad Zakir Hossain et al.
Bioscience, biotechnology, and biochemistry, 66(10), 2068-2076 (2002-11-27)
A major species of glutathione S-transferase (GST), Pugf, was highly purified from pumpkin flowers. Two-dimensional electrophoresis of the purified enzyme gave two adjacent protein spots. The specific activity of the purified enzyme was 2.4 micromol min(-1) mg(-1) protein for 1-chloro-2,4-dinitrobenzene.
Mohammad Anwar Hossain et al.
Bioscience, biotechnology, and biochemistry, 73(9), 2007-2013 (2009-09-08)
Glyoxalase I was highly purified from onion bulbs by DEAE-cellulose, hydroxyapatite, and S-hexylglutathione-agarose column chromatography. With 356 micromol min(-1) mg(-1) protein, the specific enzymatic activity of the purified enzyme is the highest reported to date in plants. The purified enzyme
T Primiano et al.
Cancer research, 55(19), 4319-4324 (1995-10-01)
Oltipraz [5-(2-pyrazinyl)-4-methyl-1,2-dithiole-3-thione] protects against chemical carcinogenesis in several animal models and is currently under evaluation as a possible chemopreventive agent in humans. Ideally, clinical chemopreventive interventions use dosing regimens that maximize efficacy while minimizing toxicity. Toward this end, the chemopreventive
A Gregory Bruce et al.
Virology journal, 6, 205-205 (2009-11-20)
ORF59 DNA polymerase processivity factor of the human rhadinovirus, Kaposi's sarcoma-associated herpesvirus (KSHV), is required for efficient copying of the genome during virus replication. KSHV ORF59 is antigenic in the infected host and is used as a marker for virus

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service