H6758
L-Histidyldiazobenzylphosphonic acid-Agarose
saline suspension
About This Item
Recommended Products
form
saline suspension
extent of labeling
≥6 μmol per mL
matrix
Cross-linked 4% beaded agarose
matrix activation
cyanogen bromide
matrix attachment
α-amino
matrix spacer
1 atom
storage temp.
2-8°C
Application
Physical form
Disclaimer

Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service