This specially prepared, chemically modified form of KLH is highly water-soluble and provides a large number of available carboxyl groups for conjugation to hapten. The conjugation of succinylated KLH to an amine group-containing hapten (e.g., peptide), mediated by water-soluble carbodiimide as cross-linking reagent, proceeds favorably between the newly introduced carboxyl groups of KLH and the amine groups of the hapten.
Physical form
Lyophilized with stabilizing buffer
Preparation Note
Prepared by modification of KLH with succinic anhydride.
Reconstitution
Reconstitute with 2 mL deionized water to yield an opalescent solution of KLH at 5 mg/ml in sodium phosphate buffer, pH 7.4, containing 0.32 M NaCl and 20 mM sucrose. Reconstituted protein solution may be stored up to two months at –20°C.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Journal of animal science, 84(4), 843-852 (2006-03-18)
Dietary guar gum and cellulose were studied as purified soluble and insoluble nonstarch polysaccharide (NSP) sources, respectively. A control diet containing 14% cornstarch was formulated. A 7% guar gum, a 7% cellulose, and a 7% guar gum + 7% cellulose
Hersckovits, T.
Comparative Biochemistry and Physiology, 91B, 579-579 (1988)
Journal of immunological methods, 156(2), 137-149 (1992-12-08)
In this report, we have described methods to improve the efficiency of coupling synthetic peptides to keyhole limpet hemocyanin (KLH) and for the analysis of the composition of the resulting peptide-protein conjugates. KLH was first dissolved in buffers containing 3
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.