Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GERPN1520S

Hybond®-XL hybridization membranes

W × L 15 cm × 20 cm, sheets, pack of 10 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
NACRES:
NB.22

material

positively charged nylon
sheets

form

solid

packaging

pack of 10 ea

manufacturer/tradename

Cytiva RPN1520S

W × L

15 cm × 20 cm

color

White

pore size

0.45 μm pore size

Looking for similar products? Visit Product Comparison Guide

Legal Information

Hybond is a registered trademark of Cytiva

Storage Class

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service