Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GEPA15621

Sigma-Aldrich

Cy®5.5 Hydrazide

Cytiva PA15621, pack of 1 mg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

packaging

pack of 1 mg

manufacturer/tradename

Cytiva PA15621

color

Blue (dark)

storage temp.

2-8°C

General description

Cy5.5 Hydrazide.

Application

Hydrazide esters provide a convenient method for the labeling of free carbonyl groups of glycoProteins and carbohydrates. For maximum convenience, stable active hydrazide esters are supplied and stored as solid material.

Features and Benefits

  • For the labeling of free carbonyl groups of glycoProteins and carbohydrates.
  • Range of bulk packs available for greatest economy.
  • Example protocols included with each dye pack.

Storage and Stability

Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Cy is a registered trademark of Cytiva

pictograms

Exclamation mark

signalword

Danger

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service