Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GENAMP100

Amplify

Cytiva NAMP100, pack of 1 L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.22

form

liquid

feature

wetted part: no

packaging

pack of 1 L

manufacturer/tradename

Cytiva NAMP100

storage condition

room temperature

color

brown-green
Green / Brown / Pale colour

pH

3.5-4.5

solubility

H2O: soluble
cold water: soluble (Easily soluble in cold and hot water)

General description

Amplify Fluorographic Reagent can increase sensitivity of detection and significantly reduce exposure times required for weak beta emitters.

Features and Benefits

Amersham amplify fluorographic reagent
  • Can be used with acrylamide or agarose gels with no change in protocols.
  • Odorless, inorganic and fully compatible with Sensitize Preflash unit and Hyperfilm MP.
  • Fast and convenient fluorographic reagent for 35S-, 14C- and 3H-labeled samples in gels.
  • Less than 2 h total preparation time.
  • Increased detection efficiency 1000-fold for 3H and 15-fold for 35S and 14C.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

pictograms

Exclamation mark

signalword

Danger

hcodes

Storage Class

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service