Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE80-6501-42

Nuclease Mix

Cytiva 80-6501-42

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111000
NACRES:
NA.56

feature

wetted part: no

packaging

pkg of 0.5 mL

manufacturer/tradename

Cytiva 80-6501-42

shipped in

dry ice

storage temp.

−20°C

General description

The nuclease mix offers an effective mix of DNase and RNase enzymes, as well as the necessary cofactors for optimal nuclease activity.

Application

Removal of nucleic acids is often required to avoid contamination and subsequent artifacts on 2-D gels. The Nuclease Mix offers an effective mix of DNase and RNase enzymes, as well as the necessary cofactors for optimal nuclease activity.

Features and Benefits

  • Specifically developed for removal of nucleic acids in sample preparation for IEF/2-D electrophoresis applications in proteomic studies
  • Compatible with Protease Inhibitor Mix
  • Ready-to-use: Simply add the mix to your sample and incubate.
  • Does not contain EDTA, which is an inhibitor of nuclease.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

hcodes

Storage Class

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service