Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE27-9254-01

Sigma-Aldrich

QuickPrep mRNA Purification Kit

Cytiva 27-9254-01, pack of 1 kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

packaging

pack of 1 kit

manufacturer/tradename

Cytiva 27-9254-01

capacity

25 μg binding capacity(poly(A) RNA)

shipped in

wet ice

storage temp.

2-8°C

General description

QuickPrep Micro mRNA Purification Kit provides sufficient reagents for 24 purifications, making it an excellent choice for purifying mRNA from a large number of samples.

application

The two illustra QuickPrep kits differ with respect to the number of purifications, the maximum amount of starting sample that can be used, the time required to purify mRNA, and the prepackaging format of the kit components. Neither kit is applicable for mRNAs without poly(A+) tails (e.g., bacteria). The components in each kit are listed in the table below

Features and Benefits

  • Fast and easy to use with no need for intermediate purification of total RNA
  • Preserve integrity of mRNA by utilizing guanidine thiocyanate during crucial early stages of purification
  • Utilize speed and selectivity of oligo(dT)-cellulose spin column chromatography to purify high-quality mRNA in yields suitable for direct use in cDNA synthesis and subsequent PCR, Northern blot hybridization, and in vitro translation.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

QuickPrep is a trademark of Cytiva
illustra is a trademark of Cytiva

pictograms

Exclamation mark

signalword

Warning

Storage Class

"" - ""


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service