Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-5172-01

Superose® 6 10/300 GL

Cytiva 17-5172-01, column L × I.D. 30 cm × 10 mm, 13 μm avg. part. size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

material

borosilicate glass column
polypropylene filter

mol wt

5000-5,000,000 Da (globular proteins)

packaging

pkg of 1 ea

manufacturer/tradename

Cytiva 17-5172-01

parameter

0.3-0.5 mL/min flow rate
1.5 Mpa (217 psi)
75-500 mL sample volume

bed volume

24 mL

column L × I.D.

30 cm × 10 mm

matrix

agarose

matrix active group

agarose phase

avg. part. size

13 μm

cleaning

1-14

working range

3-12

separation technique

size exclusion (SEC)

General description

Superose® 6 10/300 GL are prepacked Tricorn columns for high-performance gel filtration with a broad protein fractionation range of 5 000 to 5 000 000.

Features and Benefits

  • Achieve high-resolution separations across exceptionally broad molecular-weight ranges.
  • Easy, efficient scale-up from prepacked Superose® 10/300 GL columns to Superose® prep grade lab packs.
  • Low ionic strength eluents (< 0.05 M) can utilize a hydrophobic interaction component to alter the selectivity of Superose® for some lipids, peptides, and small aromatic compounds.
  • Prepacked Tricorn columns are durable, high-performance galss columns for reliable and reproducible separations.

Suitability

Stable in all common buffers: 1 M acetic acid, 8 M urea, 6 M guanidine hydrochloride, 1% SDS, organic solvents, 0.5 M NaOH (for cleaning-in-place)
for high-resolution separations across exceptionally broad molecular-weight ranges

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Superose is a registered trademark of Cytiva

pictograms

Flame

signalword

Warning

hcodes

Storage Class

"" - ""


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service