Recommended Products
form
solid
feature
wetted part: no
packaging
pack of 5 g
manufacturer/tradename
Cytiva 17-1318-02
color
White
pH range
0-5.1
solubility
cold water: soluble (Easily soluble in cold and hot water)
storage temp.
2-8°C
General description
Storage and Stability
Analysis Note
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
This page describes protein precipitation as an optional step in sample preparation for 2-D electrophoresis with Cytiva products.
This page describes components of rehydration solutions for use in 2-D Electrophoresis with Cytiva products.
Protocols
This page describes typical rehydration solutions for use in gel electrophoresis with Cytiva products.
This page describes two-dimensional separation of protein samples with Cytiva products.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service