Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-1314-01

CHAPS

Cytiva 17-1314-01, pack of 1 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

sterility

non-sterile

form

solid

feature

wetted part: no

packaging

pack of 1 g

manufacturer/tradename

Cytiva 17-1314-01

color

White

solubility

cold water: soluble (Partially soluble in cold and hot water)

storage temp.

2-8°C

Application

A nonionic or zwitterionic detergent is included in the sample solution to ensure complete sample solubilization and to prevent aggregation through hydrophobic interactions. CHAPS and Triton X-100 are detergents commonly used in IEF.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Storage Class

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page describes components of rehydration solutions for use in 2-D Electrophoresis with Cytiva products.

Protocols

This page describes typical rehydration solutions for use in gel electrophoresis with Cytiva products.

This page describes two-dimensional separation of protein samples with Cytiva products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service