Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-0615-01

High Molecular Weight SDS Calibration Kit

Cytiva 17-0615-01, pkg of 10 vials

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

form

solid

feature

wetted part: no

packaging

pkg of 10 vials

manufacturer/tradename

Cytiva 17-0615-01

storage condition

(Unopened vials should be s-red at 4°C)

color

Blue

solubility

cold water: soluble (Easily soluble in cold and hot water)

storage temp.

2-8°C

General description

The calibration kit is specifically designed for electrophoresis applications, providing precise standards for accurate molecular weight determination.

Application

High molecular weight-SDS (HMW-SDS) Calibration Kit for electrophoresis is a lyophilized mixture of five highly purified, well-characterized Proteins for use in molecular weight determination in the presence of the detergent sodium dodecyl sulphate (SDS). The molecular mass of the Protein under investigation is determined by comparing its electrophoretic mobility with that of the Proteins contained in the kit.

Features and Benefits

  • Determine molecular weight of uncharacterized Protein samples with precision by comparison with carefully selected standards.
  • Get clear, sharp bands for easy measurement of migration distances.
  • Bands can be visualized by using Deep Purple Total Protein Stain, Coomassie Brilliant Blue, or silver staining.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service